missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STRAP Protein

A DNA sequence encoding the Homo sapiens (Human) STRAP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8281-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16013751

  • 32325.00 SEK / 1mg
Estimated Shipment: 17-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_009109.3
STRAP Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
11171
Greater than 80% as determined by SDS-PAGE.
Research Use Only
STRAP
Recombinant Protein
E. coli
MAMRQTPLTC SGHTRPVVDL AFSGITPYGY FLISACKDGK PMLRQGDTGD WIGTFLGHKG AVWGATLNKD ATKAATAAAD FTAKVWDAVS GDELMTLAHK HIVKTVDFTQ DSNYLLTGGQ DKLLRIYDLN KPEAEPKEIS GHTSGIKKAL WCSEDKQILS ADDKTVRLWD HATMTEVKSL NFNMSVSSME YIPEGEILVI TYGRSIAFHS AVSLDPIKSF EAPATINSAS LHPEKEFLVA GGEDFKLYKY DYNSGEELES YKGHFGPIHC VRFSPDGELY ASGSEDGTLR LWQTVVGKTY GLWKCVLPEE DSGELAKPKI GFPETTEEEL EEIASENSDC IFPSAPDVKA
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers