Learn More
enQuireBio™ Recombinant Human Thioredoxin domain-containing protein 1
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Specifications
Specifications
Accession Number | NP_001139021.1 |
Gene ID (Entrez) | 81567 |
Name | Thioredoxin domain-containing protein 1 |
Quantity | 50 μg |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | TXNDC5 |
Protein Length | Full Length of Isoform 2 |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Show More |
Product Content Correction
Product Title
enQuireBio™ Recombinant Human Thioredoxin domain-containing protein 1 >
Spot an opportunity for improvement?Share a Content Correction