missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thymidine kinase Protein

A DNA sequence encoding the Homo sapiens (Human) Thymidine kinase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6799-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005940

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

YP_009137097.1
Thymidine kinase Protein
Research Use Only
TK
Recombinant Protein
E. coli
MASYPCHQHA SAFDQAARSR GHNNRRTALR PRRQQEATEV RPEQKMPTLL RVYIDGPHGM GKTTTTQLLV ALGSRDDIVY VPEPMTYWRV LGASETIANI YTTQHRLDQG EISAGDAAVV MTSAQITMGM PYAVTDAVLA PHIGGEAGSS HAPPPALTLI FDRHPIAALL CYPAARYLMG SMTPQAVLAF VALIPPTLPG TNIVLGALPE DRHIDRLAKR QRPGERLDLA MLAAIRRVYG LLANTVRYLQ CGGSWREDWG QLSGTAVPPQ GAEPQSNAGP RPHIGDTLFT LFRAPELLAP NGDLYNVFAW ALDVLAKRLR SMHVFILDYD QSPAGCRDAL LQLTSGMVQT HVTTPGSIPT ICDLARTFAR EMGEAN
Greater than 90% as determined by SDS-PAGE.
24271467
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers