missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcription factor BTF3 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcription factor BTF3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8384-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_001032726.1 | |
Transcription factor BTF3 Protein | |
Research Use Only | |
BTF3 | |
Recombinant Protein | |
E. coli | |
TIMNQEKLAK LQAQVRIGGK GTARRKKKVV HRTATADDKK LQFSLKKLGV NNISGIEEVN MFTNQGTVIH FNNPKVQASL AANTFTITGH AETKQLTEML PSILNQLGAD SLTSLRRLAE ALPKQSVDGK APLATGEDDD DEVPDLVENF DEASKNEAN | |
Greater than 90% as determined by SDS-PAGE. |
689 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Transcription factor BTF3 Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction