missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Human Transcriptional enhancer factor TEF-5 Protein Product Code.: 16012271

enQuireBio™ Recombinant Human Transcriptional enhancer factor TEF-5 Protein

Product Code. p-7154712
10 μg, 10µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16012271

Brand: enQuireBio™ QP8032ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Transcriptional enhancer factor TEF-5, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_003205.2
Gene ID (Entrez) 7005
Name Transcriptional enhancer factor TEF-5 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol TEAD3
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MNLDQVSKDK ALQSMASMSS AQIVSASVLQ NKFSPPSPLP QAVFSTSSRF WSSPPLLGQQ PGPSQDIKPF AQPAYPIQPP LPPTLSSYEP LAPLPSAAAS VPVWQDRTIA SSRLRLLEYS AFMEVQRDPD TYSKHLFVHI GQTNPAFSDP PLEAVDVRQI YDKFPEKKGG LKELYEKGPP NAFFLVKFWA DLNSTIQEGP GAFYGVSSQY SSADSMTISV STKVCSFGKQ VVEKVETEYA RLENGRFVYR IHRSPMCEYM INFIHKLKHL PEKYMMNSVL ENFTILQVVT SRDSQETLLV IAFVFEVSTS EHGAQHHVYK LVKD
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Transcriptional enhancer factor TEF-5 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.