missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tripartite motif-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Tripartite motif-containing protein 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8134-iv-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_149023.2 | |
Tripartite motif-containing protein 1 | |
Research Use Only | |
TRIM5 | |
Recombinant Protein | |
E. coli | |
MASGILVNVK EEVTCPICLE LLTQPLSLDC GHSFCQACLT ANHKKSMLDK GESSCPVCRI SYQPENIRPN RHVANIVEKL REVKLSPEGQ KVDHCARHGE KLLLFCQEDG KVICWLCERS QEHRGHHTFL TEEVAREYQV KLQAALEMLR QKQQEAEELE ADIREEKASW KTQIQYDKTN VLADFEQLRD ILDWEESNEL QNLEKEEEDI LKSLTNSETE MVQQTQSLRE LISDLEHRLQ GSVMELLQGV DGVIKRTENV TLKKPETFPK NQRRVFRAPD LKGMLEVFRE LTDVRRYWVD VTVAPNNISC AVISEDKRQV SSPKPQIIYG ARGTRYQTFV NFNYCTGILG SQSITSGKHY WEVDVSKKTA WILGVCAGFQ PDAMCNIEKN ENYQPKYGYW VIGLEEGVKC SAFQDSSFHT PSVPFIVPLS VIICPDRVGV FLDYEACTVS FFNITNHGFL IYKFSHCSFS QPVFPYLNPR KCGVPMTLCS PSS | |
Greater than 85% as determined by SDS-PAGE. |
85363 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |