missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tripartite motif-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Tripartite motif-containing protein 5, was expressed in the hosts and tags indicated.
4660.00 SEK - 19270.00 SEK
Specifications
Accession Number | NP_149023.2 |
---|---|
Gene ID (Entrez) | 85363 |
Name | Tripartite motif-containing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16062841
|
enQuireBio™
QP8134-IV-100UG |
100 μg |
19270.00 SEK
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16082841
|
enQuireBio™
QP8134-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072841
|
enQuireBio™
QP8134-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_149023.2 | |
Tripartite motif-containing protein 1 | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
85363 | |
Research Use Only | |
TRIM5 | |
Recombinant Protein | |
E. coli | |
MASGILVNVK EEVTCPICLE LLTQPLSLDC GHSFCQACLT ANHKKSMLDK GESSCPVCRI SYQPENIRPN RHVANIVEKL REVKLSPEGQ KVDHCARHGE KLLLFCQEDG KVICWLCERS QEHRGHHTFL TEEVAREYQV KLQAALEMLR QKQQEAEELE ADIREEKASW KTQIQYDKTN VLADFEQLRD ILDWEESNEL QNLEKEEEDI LKSLTNSETE MVQQTQSLRE LISDLEHRLQ GSVMELLQGV DGVIKRTENV TLKKPETFPK NQRRVFRAPD LKGMLEVFRE LTDVRRYWVD VTVAPNNISC AVISEDKRQV SSPKPQIIYG ARGTRYQTFV NFNYCTGILG SQSITSGKHY WEVDVSKKTA WILGVCAGFQ PDAMCNIEKN ENYQPKYGYW VIGLEEGVKC SAFQDSSFHT PSVPFIVPLS VIICPDRVGV FLDYEACTVS FFNITNHGFL IYKFSHCSFS QPVFPYLNPR KCGVPMTLCS PSS | |
Greater than 85% as determined by SDS-PAGE. |