missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tubulin beta-4A chain Protein
A DNA sequence encoding the Homo sapiens (Human) Tubulin beta-4A chain, was expressed in the hosts and tags indicated.
2955.00 SEK - 31480.00 SEK
Specifications
Accession Number | NP_001276052.1 |
---|---|
Gene ID (Entrez) | 10382 |
Name | Tubulin beta-4A chain Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036300
|
enQuireBio™
QP6853-ye-10ug |
10 ÎĽg | N/A | ||||||
16076300
|
enQuireBio™
QP6853-ye-50ug |
50 ÎĽg | N/A | ||||||
16026300
|
enQuireBio™
QP6853-ye-100ug |
100 ÎĽg |
10850.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16056300
|
enQuireBio™
QP6853-ye-200ug |
200 ÎĽg | N/A | ||||||
16066300
|
enQuireBio™
QP6853-ye-500ug |
500 ÎĽg |
31245.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16046300
|
enQuireBio™
QP6853-ye-1mg |
1 mg | N/A | ||||||
Specifications
NP_001276052.1 | |
Tubulin beta-4A chain Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
10382 | |
Research Use Only | |
TUBB4A | |
Recombinant Protein | |
Yeast | |
MREIVHLQAG QCGNQIGAKF WEVISDEHGI DPTGTYHGDS DLQLERINVY YNEATGGNYV PRAVLVDLEP GTMDSVRSGP FGQIFRPDNF VFGQSGAGNN WAKGHYTEGA ELVDAVLDVV RKEAESCDCL QGFQLTHSLG GGTGSGMGTL LISKIREEFP DRIMNTFSVV PSPKVSDTVV EPYNATLSVH QLVENTDETY CIDNEALYDI CFRTLKLTTP TYGDLNHLVS ATMSGVTTCL RFPGQLNADL RKLAVNMVPF PRLHFFMPGF APLTSRGSQQ YRALTVPELT QQMFDAKNMM AACDPRHGRY LTVAAVFRGR MSMKEVDEQM LSVQSKNSSY FVEWIPNNVK TAVCDIPPRG LKMAATFIGN STAIQELFKR ISEQFTAMFR RKAFLHWYTG EGMDEMEFTE AESNMNDLVS EYQQYQDATA EEGEFEEEAE EEVA | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Tubulin beta-4A chain Protein