missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Uncharacterized protein C20orf197 Protein

Product Code. 16061171
500 μg, 500µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy
This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Uncharacterized protein C20orf197, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001289742.1
Gene ID (Entrez) 284756
Name Uncharacterized protein C20orf197 Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol C20orf197
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence MVALFQHSPY WQADGYGHSH RLKCQHFKQH RQYNDKLEIS SNLGPQFNAL LNILLNIVHP TLSHDTRRSK GLKIEGLLQS RELGNSWTVT MCIWVLKALQ SSAPNKPLDW LDPMPCFQNL LARGTP
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Uncharacterized protein C20orf197 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.