missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Upstream stimulatory factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Upstream stimulatory factor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6872-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_001263302.1 | |
Upstream stimulatory factor 1 Protein | |
Research Use Only | |
USF1 | |
Recombinant Protein | |
E. coli | |
GQQKTAETEE GTVQIQEGAV ATGEDPTSVA IASIQSAATF PDPNVKYVFR TENGGQVMYR VIQVSEGQLD GQTEGTGAIS GYPATQSMTQ AVIQGAFTSD DAVDTEGTAA ETHYTYFPST AVGDGAGGTT SGSTAAVVTT QGSEALLGQA TPPGTGQFFV MMSPQEVLQG GSQRSIAPRT HPYSPKSEAP RTTRDEKRRA QHNEVERRRR DKINNWIVQL SKIIPDCSME STKSGQSKGG ILSKACDYIQ ELRQSNHRLS EELQGLDQLQ LDNDVLRQQV EDLKNKNLLL RAQLRHHGLE VVIKNDSN | |
Greater than 90% as determined by SDS-PAGE. |
7391 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Upstream stimulatory factor 1 Protein > 1mg
Spot an opportunity for improvement?Share a Content Correction