missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VPS13A Protein
A DNA sequence encoding the Homo sapiens (Human) VPS13A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7966-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_001018047.1 | |
VPS13A Protein | |
Research Use Only | |
VPS13A | |
Recombinant Protein | |
E. coli | |
RPPRFFNEDG VIRPYRLRDG TGNQMLQVME NGRFAKYKYF THVMINKTDM LMITRRGVLF VTKGTFGQLT CEWQYSFDEF TKEPFIVHGR RLRIEAKERV KSVF | |
Greater than 90% as determined by SDS-PAGE. |
23230 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human VPS13A Protein > 500μg
Spot an opportunity for improvement?Share a Content Correction