missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WD repeat-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) WD repeat-containing protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8496-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005103.2 | |
WD repeat-containing protein 1 | |
Research Use Only | |
WDR1 | |
Recombinant Protein | |
E. coli | |
SRFVNCVRFS PDGNRFATAS ADGQIYIYDG KTGEKVCALG GSKAHDGGIY AISWSPDSTH LLSASGDKTS KIWDVSVNSV VSTFPMGSTV LDQQLGCLWQ KDHLLSVSLS GYINYLDRNN PSKPLHVIKG HSKSIQCLTV HKNGGKSYIY SGSHDGHINY WDSETGENDS FAGKGHTNQV SRMTVDESGQ LISCSMDDTV RYTSLMLRDY SGQGVVKLDV QPKCVAVGPG GYAVVVCIGQ IVLLKDQRKC FSIDNPGYEP EVVAVHPGGD TVA | |
Greater than 90% as determined by SDS-PAGE. |
9948 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |