missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WD repeat-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) WD repeat-containing protein 1, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_005103.2 |
---|---|
Gene ID (Entrez) | 9948 |
Name | WD repeat-containing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16064941
|
enQuireBio™
QP8496-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16084941
|
enQuireBio™
QP8496-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16074941
|
enQuireBio™
QP8496-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094941
|
enQuireBio™
QP8496-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004951
|
enQuireBio™
QP8496-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014951
|
enQuireBio™
QP8496-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005103.2 | |
WD repeat-containing protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9948 | |
Research Use Only | |
WDR1 | |
Recombinant Protein | |
E. coli | |
SRFVNCVRFS PDGNRFATAS ADGQIYIYDG KTGEKVCALG GSKAHDGGIY AISWSPDSTH LLSASGDKTS KIWDVSVNSV VSTFPMGSTV LDQQLGCLWQ KDHLLSVSLS GYINYLDRNN PSKPLHVIKG HSKSIQCLTV HKNGGKSYIY SGSHDGHINY WDSETGENDS FAGKGHTNQV SRMTVDESGQ LISCSMDDTV RYTSLMLRDY SGQGVVKLDV QPKCVAVGPG GYAVVVCIGQ IVLLKDQRKC FSIDNPGYEP EVVAVHPGGD TVA | |
Greater than 90% as determined by SDS-PAGE. |