missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zinc finger protein 384 Protein

A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 384, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8445-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064661

  • 26990.00 SEK / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Zinc finger protein 384 Protein
Research Use Only
ZNF384
Recombinant Protein
E. coli
MEESHFNSNP YFWPSIPTVS GQIENTMFIN KMKDQLLPEK GCGLAPPHYP TLLTVPASVS LPSGISMDTE SKSDQLTPHS QASVTQNITV VPVPSTGLMT AGVSCSQRWR REGSQSRGPG LVITSPSGSL VTTASSAQTF PISAPMIVSA LPPGSQALQV VPDLSKKVAS TLTEEGGGGG GGGGSVAPKP PRGRKKKRML ESGLPEMNDP YVLSPEDDDD HQKDGKTYRC RMCSLTFYSK SEMQI
Greater than 90% as determined by SDS-PAGE.
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers