missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein 384 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 384, was expressed in the hosts and tags indicated.
1640.00 SEK - 22920.00 SEK
Specifications
Name | Zinc finger protein 384 Protein |
---|---|
Product Type | Recombinant Protein |
Protein Tag | GST |
Host Species | E. coli |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044661
|
enQuireBio™
QP8445-EC-100UG |
100 μg |
5025.00 SEK
100µg |
Estimated Shipment: 15-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
16064661
|
enQuireBio™
QP8445-EC-1MG |
1 mg |
22920.00 SEK
1mg |
Estimated Shipment: 15-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
16054661
|
enQuireBio™
QP8445-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
16074661
|
enQuireBio™
QP8445-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
16084661
|
enQuireBio™
QP8445-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
16094661
|
enQuireBio™
QP8445-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
Specifications
Zinc finger protein 384 Protein | |
GST | |
Human | |
Greater than 90% as determined by SDS-PAGE. |
Recombinant Protein | |
E. coli | |
MEESHFNSNP YFWPSIPTVS GQIENTMFIN KMKDQLLPEK GCGLAPPHYP TLLTVPASVS LPSGISMDTE SKSDQLTPHS QASVTQNITV VPVPSTGLMT AGVSCSQRWR REGSQSRGPG LVITSPSGSL VTTASSAQTF PISAPMIVSA LPPGSQALQV VPDLSKKVAS TLTEEGGGGG GGGGSVAPKP PRGRKKKRML ESGLPEMNDP YVLSPEDDDD HQKDGKTYRC RMCSLTFYSK SEMQI | |
ZNF384 |