missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Influenza A H1N1 (New Jersey / 8 / 1976) NucleoProtein Product Code.: 16057420

enQuireBio™ Recombinant Influenza A H1N1 (New Jersey / 8 / 1976) NucleoProtein

Product Code. 16057420
500 μg, 500µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16057420

Brand: enQuireBio™ QP7117500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Influenza A virus Influenza A H1N1 (strain A/New Jersey/8/1976) Nucleoprotein, was expressed in the hosts and tags indicated.

Specifications

Name Influenza A H1N1 (New Jersey / 8 / 1976) NucleoProtein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol NP
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Influenza
Species E. coli
Protein Tag His-SUMO
Sequence MASQGTKRSY EQMETGGERQ DATEIRASVG RMIGGIGRFY IQMCTELKLS DYEGRLIQNS ITIERMVLSA FDERRNKYLE EHPSAGKDPK KTGGPIYKRI DGKWMRELIL YDKEEIRRVW RQANNGEDAT AGLTHIMIWH SNLNDATYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTIA MELIGMIKRG INDRNFWRGE NGRRTRIAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGLAVASGHD FEREGYSLVG IDPFKLLQNS QVFSLIRPNE NPAHKSQLVW MACHSAAFED LRVSGFIRGK KVVPRGRLST RGVQIASNEN VEAMDSSTLE LRSRYWAIRT RSGGNTNQQK ASADQISVQP TFSVQRNLPF ERATVMAAFI GDNEGRTSDM RTEIIRMMES AKPEDLSFQG RGVFELSDEK ATNPIVPSFD MNNEGSYFFG DNAEEYDN
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Influenza A H1N1 (New Jersey / 8 / 1976) NucleoProtein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.