missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H3N2 (Port Chalmers / 1 / 1973) NucleoProtein
A DNA sequence encoding the Influenza A virus (strain A/Port Chalmers/1/1973 H3N2) Influenza A H3N2 (strain A/Port Chalmers/1/1973) Nucleoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7624-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza A H3N2 (Port Chalmers / 1 / 1973) NucleoProtein | |
Research Use Only | |
NP | |
Recombinant Protein | |
E. coli | |
MASQGTKRSY EQMETDGERQ NATEIRASVG KMIDGIGRFY IQMCTELKLS DYEGRLIQNS LTIERMVLSA FDERRNRYLE EHPSAGKDPK KTGGPIYKRV DGKWMRELVL YDKEEIRRIW RQANNGDDAT AGLTHMMIWH SNLNDTTYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTMV MELIRMIKRG INDRNFWRGE NGRKTRGAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGPAVASGYN FEKEGYSLVG IDPFKLLQNS QVYSLIRPNE NPAHKSQLVW MACNSAAFED LRLLSFIRGT KVSPRGKLST RGVQIASNEN MDTMESSTLE LRSRYWAIRT RSGGNTNQQR ASAGQISVQP AFSVQRNLPF DKSTIMAAFT GNTEGRTSDM RAEIIRMMEG AKPEEVSFRG RGVFELSDEK ATNPIVPSFD MSNEGSYFFG DNAEEYDN | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |