missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H3N2 (Port Chalmers / 1 / 1973) NucleoProtein
A DNA sequence encoding the Influenza A virus (strain A/Port Chalmers/1/1973 H3N2) Influenza A H3N2 (strain A/Port Chalmers/1/1973) Nucleoprotein, was expressed in the hosts and tags indicated.
3180.00 SEK - 38570.00 SEK
Specifications
Name | Influenza A H3N2 (Port Chalmers / 1 / 1973) NucleoProtein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | NP |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059860
|
enQuireBio™
QP7624-EC-100UG |
100 μg |
11125.00 SEK
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16079860
|
enQuireBio™
QP7624-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16069860
|
enQuireBio™
QP7624-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089860
|
enQuireBio™
QP7624-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099860
|
enQuireBio™
QP7624-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009870
|
enQuireBio™
QP7624-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Influenza A H3N2 (Port Chalmers / 1 / 1973) NucleoProtein | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
NP | |
Recombinant Protein | |
E. coli | |
MASQGTKRSY EQMETDGERQ NATEIRASVG KMIDGIGRFY IQMCTELKLS DYEGRLIQNS LTIERMVLSA FDERRNRYLE EHPSAGKDPK KTGGPIYKRV DGKWMRELVL YDKEEIRRIW RQANNGDDAT AGLTHMMIWH SNLNDTTYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTMV MELIRMIKRG INDRNFWRGE NGRKTRGAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGPAVASGYN FEKEGYSLVG IDPFKLLQNS QVYSLIRPNE NPAHKSQLVW MACNSAAFED LRLLSFIRGT KVSPRGKLST RGVQIASNEN MDTMESSTLE LRSRYWAIRT RSGGNTNQQR ASAGQISVQP AFSVQRNLPF DKSTIMAAFT GNTEGRTSDM RAEIIRMMEG AKPEEVSFRG RGVFELSDEK ATNPIVPSFD MSNEGSYFFG DNAEEYDN | |
Greater than 90% as determined by SDS-PAGE. |