missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL2 / Interleukin-2 Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL2 / Interleukin-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6217-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001040595.1 | |
IL2 / Interleukin-2 Protein | |
10 μg | |
Not determined. | |
Full Length of Mature Protein | |
Macaque | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
708017 | |
Greater than 82.1% as determined by SDS-PAGE. | |
Research Use Only | |
IL2 | |
Recombinant Protein | |
E. coli | |
APTSSSTKKT QLQLEHLLLD LQMILNGINN YKNPKLTRML TFKFYMPKKA TELKHLQCLE EELKPLEEVL NLAQSKNFHL RDTKDLISNI NVIVLELKGS ETTLMCEYAD ETATIVEFLN RWITFCQSII STLT |