missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ADH7 Protein
A DNA sequence encoding the Mus musculus (Mouse) ADH7, was expressed in the hosts and tags indicated.
3180.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_033756.2 |
---|---|
Gene ID (Entrez) | 11529 |
Name | ADH7 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16090221
|
enQuireBio™
QP7693-ec-10ug |
10 ÎĽg | N/A | ||||||
16030231
|
enQuireBio™
QP7693-ec-50ug |
50 ÎĽg | N/A | ||||||
16080221
|
enQuireBio™
QP7693-ec-100ug |
100 ÎĽg |
11590.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16010231
|
enQuireBio™
QP7693-ec-200ug |
200 ÎĽg | N/A | ||||||
16020231
|
enQuireBio™
QP7693-ec-500ug |
500 ÎĽg | N/A | ||||||
16000231
|
enQuireBio™
QP7693-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_033756.2 | |
ADH7 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
11529 | |
Research Use Only | |
ADH7 | |
Recombinant Protein | |
E. coli | |
MGTAGKVIKC KAAVLWGVNQ PFSIEEIEVA PPKAKEVRVK ILATGICRTD DHIIKGSMVS KFPVIVGHEA VGVVESVGEG VTTVRPGDKV IPLFLPQCRE CNACLNPEGN LCIRSDLTGR GVLADGTTRF TCKGKPVQHF MNTSTFTEYT VLDESSVAKV DGAAPPEKAC LIGCGFSTGY GAAVKTAKVT PGSTCVVFGL GGVGLSVIMG CKAAGASRII GIDINKDKFQ KALAVGATEC ISPKDSTKPI SEVLSDMTGN TIQYTFEVIG RLETMVDALS SCHMNYGTSV VVGAPPSAKM LTYDPMLLFT GRTWKGCVFG GWKSRDDVPK LVTEFLEKKF DLDQLITHTL PFNNINEGFE LLYSGKSIRT VLTF | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse ADH7 Protein