missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Mouse Alkaline Phosphatase / ALPL Protein Product Code.: 16036981

enQuireBio™ Recombinant Mouse Alkaline Phosphatase / ALPL Protein

Product Code. 16036981
1 mg, 1mg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16036981

Brand: enQuireBio™ QP8885ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) Alkaline Phosphatase / ALPL, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001274101.1
Gene ID (Entrez) 11647
Name Alkaline Phosphatase / ALPL Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Alpl
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag His
Sequence PEKERDPSYW RQQAQETLKN ALKLQKLNTN VAKNVIMFLG DGMGVSTVTA ARILKGQLHH NTGEETRLEM DKFPFVALSK TYNTNAQVPD SAGTATAYLC GVKANEGTVG VSAATERTRC NTTQGNEVTS ILRWAKDAGK SVGIVTTTRV NHATPSAAYA HSADRDWYSD NEMPPEALSQ GCKDIAYQLM HNIKDIDVIM GGGRKYMYPK NRTDVEYELD EKARGTRLDG LDLISIWKSF KPRHKHSHYV WNRTELLALD PSRVDYLLGL FEPGDMQYEL NRNNLTDPSL SEMVEVALRI LTKNLKGFFL LVEGGRIDHG HHEGKAKQAL HEAVEMDQAI GKAGAMTSQK DTLTVVTADH SHVFTFGGYT PRGNSIFGLA PMVSDTDKKP FTAILYGNGP GYKVVDGERE NVSMVDYAHN NYQAQSAVPL RHETHGGEDV AVFAKGPMAH LLHGVHEQNY IPHVMAYASC IGANLDHCAW AGSG
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Alkaline Phosphatase / ALPL Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.