missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Apolipoprotein C-III Protein
A DNA sequence encoding the Mus musculus (Mouse) Apolipoprotein C-III, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5667-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_001276685.1 | |
Apolipoprotein C-III Protein | |
Research Use Only | |
APOC3 | |
Recombinant Protein | |
E. coli | |
EEVEGSLLLG SVQGYMEQAS KTVQDALSSV QESDIAVVAR GWMDNHFRFL KGYWSKFTDK FTGFWDSNPE DQPTPAIES | |
Greater than 90% as determined by SDS-PAGE. |
11814 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Apolipoprotein C-III Protein > 100μg
Spot an opportunity for improvement?Share a Content Correction