missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Mouse BAG family molecular chaperone regulator 3 Protein Product Code.: 16062491

enQuireBio™ Recombinant Mouse BAG family molecular chaperone regulator 3 Protein

Product Code. 16062491
500 μg, 500µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16062491

Brand: enQuireBio™ QP8071ec500ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) BAG family molecular chaperone regulator 3, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_038891.4
Gene ID (Entrez) 29810
Name BAG family molecular chaperone regulator 3 Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Bag3
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag His-SUMO
Sequence SAATQSPMMQ MASGNGASDR DPLPPGWEIK IDPQTGWPFF VDHNSRTTTW NDPRVPPEGP KDTASSANGP SRDGSRLLPI REGHPIYPQL RPGYIPIPVL HEGSENRQPH LFHAYSQPGV QRFRTEAAAA TPQRSQSPLR GGMTEAAQTD KQCGQMPATA TTAAAQPPTA HGPERSQSPA ASDCSSSSSS ASLPSSGRSS LGSHQLPRGY IPIPVIHEQN ITRPAAQPSF HQAQKTHYPA QQGEYQPQQP VYHKIQGDDW EPRPLRAASP FRSPVRGASS REGSPARSGT PVHCPSPIRV HTVVDRPQPM THREPPPVTQ PENKPESKPG PAGPDLPPGH IPIQVIRREA DSKPVSQKSP PPAEKVEVKV SSAPIPCPSP SPAPSAVPSP PKNVAAEQKA APSPAPAEPA APKSGEAETP PKHPGVLKVE AILEKVQGLE QAVDSFEGKK TDKKYLMIEE YLTKELLALD SVDPEGRADV RQARRDGVRK VQTILEKLEQ KAIDVPGQVQ VYELQPSNLE AEQPLQEIMG AVVADKDKKG PENKDPQTES QQLEAKAATP PNPSNPADSA GNLVAP
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse BAG family molecular chaperone regulator 3 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.