missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse bFGF / FGF2 Protein

A DNA sequence encoding the Mus musculus (Mouse) bFGF / FGF2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8538-ec-100ug

 View more versions of this product

Product Code. 16015171

  • 9055.00 SEK / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_032032.1
bFGF / FGF2 Protein
Research Use Only
FGF2
Recombinant Protein
E. coli
PALPEDGGAA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHV KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTEE CFFFERLESN NYNTYRSRKY SSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
Greater than 90% as determined by SDS-PAGE.
14173
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse bFGF / FGF2 Protein > 100μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.