missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Carboxylesterase 1C Protein

A DNA sequence encoding the Mus musculus (Mouse) Carboxylesterase 1C, was expressed in the hosts and tags indicated.

2430.00 SEK - 40175.00 SEK

Specifications

Accession Number NP_031980.2
Gene ID (Entrez) 13884
Name Carboxylesterase 1C Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16097610
View Documents
enQuireBio™
QP7188-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16037620
View Documents
enQuireBio™
QP7188-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16087610
View Documents
enQuireBio™
QP7188-ec-100ug
100 ÎĽg
9055.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16017620
View Documents
enQuireBio™
QP7188-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16027620
View Documents
enQuireBio™
QP7188-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16007620
View Documents
enQuireBio™
QP7188-ec-1mg
1 mg
40175.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_031980.2
Carboxylesterase 1C Protein
Not determined.
Partial (see sequence information for more details).
Mouse
GST
Tris-based buffer, 50% glycerol
13884
Research Use Only
Ces1c
Recombinant Protein
E. coli
HSLLPPVVDT TQGKVLGKYI SLEGFEQPVA VFLGVPFAKP PLGSLRFAPP QPAEPWSFVK NATSYPPMCS QDAGWAKILS DMFSTEKEIL PLKISEDCLY LNIYSPADLT KSSQLPVMVW IHGGGLVIGG ASPYNGLALS AHENVVVVTI QYRLGIWGLF STGDEHSPGN WAHLDQLAAL RWVQDNIANF GGNPDSVTIF GESSGGISVS VLVLSPLGKD LFHRAISESG VVINTNVGKK NIQAVNEIIA TLSQCNDTSS AAMVQCLRQK TESELLEISG KLVQYNISLS TMIDGVVLPK APEEILAEKS FNTVPYIVGF NKQEFGWIIP MMLQNLLPEG KMNEETASLL LRRFHSELNI SESMIPAVIE QYLRGVDDPA KKSELILDMF GDIFFGIPAV LLSRSLRDAG VSTYMYEFRY RPSFVSDKRP QTVEGDHGDE IFFVFGAPLL KEGASEEETN LSKMVMKFWA NFARNGNPNG EGLPHWPEYD EQEGYLQIGA TTQQAQRLKA EEVAFWTELL AKNPPETDPT EH
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Carboxylesterase 1C Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.