Learn More
enQuireBio™ Recombinant Mouse CD3e / CD3 epsilon Protein
A DNA sequence encoding the Mus musculus (Mouse) CD3e / CD3 epsilon, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9045-ye-1mg
This item is not returnable.
View return policy
Specifications
NP_031674.1 | |
CD3e / CD3 epsilon Protein | |
Research Use Only | |
CD3E | |
Recombinant Protein | |
Yeast | |
DAENIEYKVS ISGTSVELTC PLDSDENLKW EKNGQELPQK HDKHLVLQDF SEVEDSGYYV CYTPASNKNT YLYLKARVCE YCVEVD | |
Greater than 90% as determined by SDS-PAGE. |
12501 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Product Title
enQuireBio™ Recombinant Mouse CD3e / CD3 epsilon Protein > 1mg
Spot an opportunity for improvement?Share a Content Correction