missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD3e / CD3 epsilon Protein

A DNA sequence encoding the Mus musculus (Mouse) CD3e / CD3 epsilon, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9045-ye-50ug

 View more versions of this product

Product Code. 16067521

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_031674.1
CD3e / CD3 epsilon Protein
Research Use Only
CD3E
Recombinant Protein
Yeast
DAENIEYKVS ISGTSVELTC PLDSDENLKW EKNGQELPQK HDKHLVLQDF SEVEDSGYYV CYTPASNKNT YLYLKARVCE YCVEVD
Greater than 90% as determined by SDS-PAGE.
12501
50 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse CD3e / CD3 epsilon Protein > 50μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.