missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Mouse Collagenase 3 Protein Product Code.: 16057091

enQuireBio™ Recombinant Mouse Collagenase 3 Protein

Product Code. 16057091
100 μg, 100µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16057091

Brand: enQuireBio™ QP8902ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) Collagenase 3, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_032633.1
Gene ID (Entrez) 17386
Name Collagenase 3 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol MMP13
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag His
Sequence YNVFPRTLKW SQTNLTYRIV NYTPDMSHSE VEKAFRKAFK VWSDVTPLNF TRIYDGTADI MISFGTKEHG DFYPFDGPSG LLAHAFPPGP NYGGDAHFDD DETWTSSSKG YNLFIVAAHE LGHSLGLDHS KDPGALMFPI YTYTGKSHFM LPDDDVQGIQ FLYGPGDEDP NPKHPKTPEK CDPALSLDAI TSLRGETMIF KDRFFWRLHP QQVEAELFLT KSFWPELPNH VDAAYEHPSR DLMFIFRGRK FWALNGYDIL EGYPRKISDL GFPKEVKRLS AAVHFENTGK TLFFSENHVW SYDDVNQTMD KDYPRLIEEE FPGIGNKVDA VYEKNGYIYF FNGPIQFEYS IWSNRIVRVM PTNSILWC
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Collagenase 3 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.