missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL4 / PF4 Protein
A DNA sequence encoding the Mus musculus (Mouse) CXCL4 / PF4 Protein, was expressed in the hosts and tags indicated.
2515.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_064316.1 |
---|---|
Gene ID (Entrez) | 56744 |
Name | CXCL4 / PF4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16093630
|
enQuireBio™
QP6484-ec-10ug |
10 ÎĽg | N/A | ||||||
16033640
|
enQuireBio™
QP6484-ec-50ug |
50 ÎĽg | N/A | ||||||
16083630
|
enQuireBio™
QP6484-ec-100ug |
100 ÎĽg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16013640
|
enQuireBio™
QP6484-ec-200ug |
200 ÎĽg | N/A | ||||||
16023640
|
enQuireBio™
QP6484-ec-500ug |
500 ÎĽg | N/A | ||||||
16003640
|
enQuireBio™
QP6484-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_064316.1 | |
CXCL4 / PF4 Protein | |
Not determined. | |
Full Length of Mature Protein | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
56744 | |
Research Use Only | |
Pf4 | |
Recombinant Protein | |
E. coli | |
VTSAGPEESD GDLSCVCVKT ISSGIHLKHI TSLEVIKAGR HCAVPQLIAT LKNGRKICLD RQAPLYKKVI KKILES | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse CXCL4 / PF4 Protein