missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Cytochrome P450 2E1 Protein

Product Code. 16007641
1 mg, 1mg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy
This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) Cytochrome P450 2E1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_067257.1
Gene ID (Entrez) 13106
Name Cytochrome P450 2E1 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Cyp2e1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Mouse
Species Yeast
Protein Tag His
Sequence AVLGITVALL VWIATLLLVS IWKQIYRSWN LPPGPFPIPF FGNIFQLDLK DIPKSLTKLA KRFGPVFTLH LGQRRIVVLH GYKAVKEVLL NHKNEFSGRG DIPVFQEYKN KGIIFNNGPT WKDVRRFSLS ILRDWGMGKQ GNEARIQREA HFLVEELKKT KGQPFDPTFL IGCAPCNVIA DILFNKRFDY DDKKCLELMS LFNENFYLLS TPWIQAYNYF SDYLQYLPGS HRKVMKNVSE IRQYTLGKAK EHLKSLDINC PRDVTDCLLI EMEKEKHSQE PMYTMENISV TLADLFFAGT ETTSTTLRYG LLILMKYPEI EEKLHEEIDR VIGPSRAPAV RDRMNMPYMD AVVHEIQRFI NLVPSNLPHE ATRDTVFRGY VIPKGTVVIP TLDSLLFDNY EFPDPETFKP EHFLNENGKF KYSDYFKAFS AGKRVCVGEG LARMELFLLL SAILQHFNLK SLVDPKDIDL SPVTIGFGSI PREFKLCVIP RS
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Cytochrome P450 2E1 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.