missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse EF-hand domain-containing protein D2 Protein

A DNA sequence encoding the Mus musculus (Mouse) EF-hand domain-containing protein D2, was expressed in the hosts and tags indicated.

2515.00 SEK - 40175.00 SEK

Specifications

Name EF-hand domain-containing protein D2 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Efhd2
Protein Length Full Length
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16012361
View Documents
enQuireBio™
QP8046-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16052361
View Documents
enQuireBio™
QP8046-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16002361
View Documents
enQuireBio™
QP8046-ec-100ug
100 ÎĽg
9055.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16032361
View Documents
enQuireBio™
QP8046-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16042361
View Documents
enQuireBio™
QP8046-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16022361
View Documents
enQuireBio™
QP8046-ec-1mg
1 mg
40175.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

EF-hand domain-containing protein D2 Protein
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Research Use Only
Efhd2
Recombinant Protein
E. coli
ATDELASKLS RRLQMEGEGG EATEQPGLNG AAAAAAAEAP DETAQALGSA DDELSAKLLR RADLNQGIGE PQSPSRRVFN PYTEFKEFSR KQIKDMEKMF KQYDAGRDGF IDLMELKLMM EKLGAPQTHL GLKSMIQEVD EDFDSKLSFR EFLLIFRKAA AGELQEDSGL QVLARLSEID VSTEGVKGAK NFFEAKVQAI NVSSRFEEEI KAEQEERKKQ AEEVKQRKAA FKELQSTFK
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse EF-hand domain-containing protein D2 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.