missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Galectin-6 Protein
A DNA sequence encoding the Mus musculus (Mouse) Galectin-6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6298-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_034837.2 | |
Galectin-6 Protein | |
Research Use Only | |
Lgals6 | |
Recombinant Protein | |
E. coli | |
MAYVPAPGYQ PTYNPTLPYK RPIPGGLSVG MSFYIQGTAK ENMRRFHVNF AVGQDDGADV AFHFNPRFDG WDKVVFNTKQ SGRWGKEEEK SMPFQKGKHF ELVFMVMPEH YKVVVNGSPF YEYGHRLPVQ MVTHLQVDGD LELQSINFFG VQPAETKYPA MTGPPVFNPC LPYVGALQGG FTVRRTIIIK GYVLPTAKTF AINFRVGSSE DIALHINPRI GDCLVRNSYM NGSWGTEERM VAYNPFGPGQ FFDLSIRCGM DRFKVFANGI HLFNFSHRFQ ALRKINTLEI NGDLTLSYVH I | |
Greater than 90% as determined by SDS-PAGE. |
16857 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |