missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Galectin-7 Protein

A DNA sequence encoding the Mus musculus (Mouse) Galectin-7, was expressed in the hosts and tags indicated.

2515.00 SEK - 26260.00 SEK

Specifications

Name Galectin-7 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol LGALS7
Protein Length Full Length
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16072280
View Documents
enQuireBio™
QP6299-ye-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16012290
View Documents
enQuireBio™
QP6299-ye-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16062280
View Documents
enQuireBio™
QP6299-ye-100ug
100 ÎĽg
9055.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16092280
View Documents
enQuireBio™
QP6299-ye-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16002290
View Documents
enQuireBio™
QP6299-ye-500ug
500 ÎĽg
25885.00 SEK
500µg
Please to purchase this item. Need a web account? Register with us today!
16082280
View Documents
enQuireBio™
QP6299-ye-1mg
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

Galectin-7 Protein
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Research Use Only
LGALS7
Recombinant Protein
Yeast
SATHHKTSLP QGVRVGTVMR IRGLVPDQAG RFHVNLLCGE EQGADAALHF NPRLDTSEVV FNTKQQGKWG REERGTGIPF QRGQPFEVLL IATEEGFKAV VGDDEYLHFH HRLPPARVRL VEVGGDVQLH SLNIF
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Galectin-7 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.