missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Glucose-6-phosphate isomerase Protein

A DNA sequence encoding the Mus musculus (Mouse) Glucose-6-phosphate isomerase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9161-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16057751

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_032181.2
Glucose-6-phosphate isomerase Protein
10 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
14751
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Gpi
Recombinant Protein
Yeast
AALTRNPQFQ KLLEWHRANS ANLKLRELFE ADPERFNNFS LNLNTNHGHI LVDYSKNLVN KEVMQMLVEL AKSRGVEAAR DNMFSGSKIN YTENRAVLHV ALRNRSNTPI KVDGKDVMPE VNRVLDKMKS FCQRVRSGDW KGYTGKSITD IINIGIGGSD LGPLMVTEAL KPYSKGGPRV WFVSNIDGTH IAKTLASLSP ETSLFIIASK TFTTQETITN AETAKEWFLE AAKDPSAVAK HFVALSTNTA KVKEFGIDPQ NMFEFWDWVG GRYSLWSAIG LSIALHVGFD HFEQLLSGAH WMDQHFLKTP LEKNAPVLLA LLGIWYINCY GCETHALLPY DQYMHRFAAY FQQGDMESNG KYITKSGARV DHQTGPIVWG EPGTNGQHAF YQLIHQGTKM IPCDFLIPVQ TQHPIRKGLH HKILLANFLA QTEALMKGKL PEEARKELQA AGKSPEDLEK LLPHKVFEGN RPTNSIVFTK LTPFILGALI AMYEHKIFVQ GIMWDINSFD QWGVELGKQL AKKIEPELEG SSAVTSHDSS TNGLISFIKQ QRDTKLE
Videos
SDS
Documents