missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Mouse Glutathione S-transferase P 1 Protein Product Code.: 16011000

enQuireBio™ Recombinant Mouse Glutathione S-transferase P 1 Protein

Product Code. 16011000
1 mg, 1mg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16011000

Brand: enQuireBio™ QP6126ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) Glutathione S-transferase P 1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_038569.1
Gene ID (Entrez) 14870
Name Glutathione S-transferase P 1 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Gstp1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag His-SUMO
Sequence PPYTIVYFPV RGRCEAMRML LADQGQSWKE EVVTIDTWMQ GLLKPTCLYG QLPKFEDGDL TLYQSNAILR HLGRSLGLYG KNQREAAQMD MVNDGVEDLR GKYVTLIYTN YENGKNDYVK ALPGHLKPFE TLLSQNQGGK AFIVGDQISF ADYNLLDLLL IHQVLAPGCL DNFPLLSAYV ARLSARPKIK AFLSSPEHVN RPINGNGKQ
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Glutathione S-transferase P 1 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.