missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Mouse Guanylate cyclase activator 2B Protein Product Code.: 16061050

enQuireBio™ Recombinant Mouse Guanylate cyclase activator 2B Protein

Code produit. 16061050
200 μg, 200µg
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
missing translation for 'unitSize'
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16061050

Brand: enQuireBio™ QP6134ec200ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) Guanylate cyclase activator 2B, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_032217.2
Gene ID (Entrez) 14916
Name Guanylate cyclase activator 2B Protein
Quantity 200 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Guca2b
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag GST
Sequence VYIKYHGFQV QLESVKKLNE LEEKEMSNPQ PRRSGLLPAV CHNPALPLDL QPVCASQEAA STFKALRTIA TDECELCINV ACTGC
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Guanylate cyclase activator 2B Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.