missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Integrin alpha-L Protein

A DNA sequence encoding the Mus musculus (Mouse) Integrin alpha-L, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6240-ec-100ug

 View more versions of this product

Product Code. 16011850

  • 9055.00 SEK / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Integrin alpha-L Protein
Research Use Only
Itgal
Recombinant Protein
E. coli
DLVFLFDGSQ SLDRKDFEKI LEFMKDVMRK LSNTSYQFAA VQFSTDCRTE FTFLDYVKQN KNPDVLLGSV QPMFLLTNTF RAINYVVAHV FKEESGARPD ATKVLVIITD GEASDKGNIS AAHDITRYII GIGKHFVSVQ KQKTLHIFAS EPVEEFVKIL DTFEKLKDLF TDL
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Integrin alpha-L Protein > 100μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.