missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Kallikrein 1-related peptidase b22 Protein
A DNA sequence encoding the Mus musculus (Mouse) Kallikrein 1-related peptidase b22, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7102-10ug
This item is not returnable.
View return policy
Specifications
NP_034244.1 | |
Kallikrein 1-related peptidase b22 Protein | |
Research Use Only | |
Klk1b22 | |
Recombinant Protein | |
E. coli | |
ILGGFKCEKN SQPWQVAVYY LDEYLCGGVL LDRNWVLTAA HCYEDKYNIW LGKNKLFQDE PSAQHRLVSK SFPHPDFNMS LLQSVPTGAD LSNDLMLLRL SKPADITDVV KPIDLPTTEP KLGSTCLASG WGSINQLIYQ NPNDLQCVSI KLHPNEVCVK AHILKVTDVM LCAGEMNGGK DTCKGDSGGP LICDGVLQGI TSWGSTPCGE PNAPAIYTKL IKFTSWIKDT MAKNP | |
Greater than 90% as determined by SDS-PAGE. |
13646 | |
10 ÎĽg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Kallikrein 1-related peptidase b22 Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction