Learn More
enQuireBio™ Recombinant Mouse OTC Protein
A DNA sequence encoding the Mus musculus (Mouse) OTC, was expressed in the hosts and tags indicated.
2430.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_032795.1 |
---|---|
Gene ID (Entrez) | 18416 |
Name | OTC Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16093430
|
enQuireBio™
QP6457-ec-10ug |
10 μg | N/A | ||||||
16033440
|
enQuireBio™
QP6457-ec-50ug |
50 μg | N/A | ||||||
16083430
|
enQuireBio™
QP6457-ec-100ug |
100 μg |
8695.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16013440
|
enQuireBio™
QP6457-ec-200ug |
200 μg | N/A | ||||||
16023440
|
enQuireBio™
QP6457-ec-500ug |
500 μg | N/A | ||||||
16003440
|
enQuireBio™
QP6457-ec-1mg |
1 mg |
38570.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_032795.1 | |
OTC Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
18416 | |
Research Use Only | |
OTC | |
Recombinant Protein | |
E. coli | |
SQVQLKGRDL LTLKNFTGEE IQYMLWLSAD LKFRIKQKGE YLPLLQGKSL GMIFEKRSTR TRLSTETGFA LLGGHPSFLT TQDIHLGVNE SLTDTARVLS SMTDAVLARV YKQSDLDTLA KEASIPIVNG LSDLYHPIQI LADYLTLQEH YGSLKGLTLS WIGDGNNILH SIMMSAAKFG MHLQAATPKG YEPDPNIVKL AEQYAKENGT KLSMTNDPLE AARGGNVLIT DTWISMGQED EKKKRLQAFQ GYQVTMKTAK VAASDWTFLH CLPRKPEEVD DEVFYSPRSL VFPEAENRKW TIMAVMVSLL TDYSPVLQKP KF | |
Greater than 90% as determined by SDS-PAGE. |