missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TPT1 / Tumor Protein
A DNA sequence encoding the Mus musculus (Mouse) TPT1 / Tumor, was expressed in the hosts and tags indicated.
2430.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_033455.1 |
---|---|
Gene ID (Entrez) | 22070 |
Name | TPT1 / Tumor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16046170
|
enQuireBio™
QP6835-ye-10ug |
10 ÎĽg | N/A | ||||||
16086170
|
enQuireBio™
QP6835-ye-50ug |
50 ÎĽg | N/A | ||||||
16036170
|
enQuireBio™
QP6835-ye-100ug |
100 ÎĽg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16066170
|
enQuireBio™
QP6835-ye-200ug |
200 ÎĽg | N/A | ||||||
16076170
|
enQuireBio™
QP6835-ye-500ug |
500 ÎĽg | N/A | ||||||
16056170
|
enQuireBio™
QP6835-ye-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_033455.1 | |
TPT1 / Tumor Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
22070 | |
Research Use Only | |
Tpt1 | |
Recombinant Protein | |
Yeast | |
MIIYRDLISH DELFSDIYKI REIADGLCLE VEGKMVSRTE GAIDDSLIGG NASAEGPEGE GTESTVVTGV DIVMNHHLQE TSFTKEAYKK YIKDYMKSLK GKLEEQKPER VKPFMTGAAE QIKHILANFN NYQFFIGENM NPDGMVALLD YREDGVTPFM IFFKDGLEME KC | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse TPT1 / Tumor Protein