missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse VEGF-C Protein
A DNA sequence encoding the Mus musculus (Mouse) VEGF-C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6880-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_033532.1 | |
VEGF-C Protein | |
Research Use Only | |
Vegfc | |
Recombinant Protein | |
E. coli | |
AHYNTEILKS IDNEWRKTQC MPREVCIDVG KEFGAATNTF FKPPCVSVYR CGGCCNSEGL QCMNTSTGYL SKTLFEITVP LSQGPKPVTI SFANHTSCRC MSKLDVYRQV HSIIRR | |
Greater than 90% as determined by SDS-PAGE. |
22341 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse VEGF-C Protein > 1mg
Spot an opportunity for improvement?Share a Content Correction