missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Mycoplasma Probable cytosol aminopeptidase Protein Product Code.: 16026830

enQuireBio™ Recombinant Mycoplasma Probable cytosol aminopeptidase Protein

Product Code. 16026830
100 μg, 100µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16026830

Brand: enQuireBio™ QP6948100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Probable cytosol aminopeptidase, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_110261.1
Gene ID (Entrez) 876829
Name Mycoplasma Probable cytosol aminopeptidase Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol pepA
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Mycoplasma
Species E. coli
Protein Tag His-SUMO
Sequence MKLNKLFDSK TVVFKKSDKY GKSDCCKTKC VEGQIEFGVK IPTDFPAFNR ALVTFLKTQK NQLNVDLDSF VELYKAENLC YKTALKVVVA SITFCETTPF TMKTEPQKNV EVAVKCDSEH TSLIKEYEVV GNYVNMARQL QDTPSDQLYP EEFVKRFEKA ATGLGVKITV LKQADLIKKK MGLLLGVNKG SEREARLLVI SYNNNKKSSE TLALVGKGIT YDSGGMNIKT GDYMRGMKYD MSGAAIVCST VLALAKNKVK TNVVAVAALT ENLPGPHAQR PDDIQTAYNG KTVEIDNTDA EGRLVLADAI SYAAKDLKAT QIIDVATLTG LMSYILSTTY TGIFSTCDMA WDAFKKAACC AGEPVWRLPM HPDYLKPLES KLADLQNSTS VKGAGSSRAA CFLAEFREGV PLIHCDIAST ASIQDLGQGV LVRTLYERAA QQAKE
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mycoplasma Probable cytosol aminopeptidase Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.