missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Shiga Like Toxin 2 Protein
A cDNA sequence encoding the Shiga Like Toxin 2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13496-500ug
Additional Details : Weight : 0.01000kg
Specifications
Shiga Like Toxin 2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Shiga Like Toxin 2 protein solution contains PBS, 50mM arginine and 0.05% sodium azide. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |