missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other T. pallidum p17 (Partial) Protein
Click to view available options
Quantity:
100 μg
1200 μg
500 μg
Unit Size:
100µg
1200µg
500µg
Specifications
Specifications
Name | T. pallidum p17 (Partial) Protein |
Quantity | 500 μg |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Species | E. coli |
Protein Tag | His |
Buffer | 70mM Tris-HCl pH 8.0, 50mM NaCl, 50% Glycerol, 1.5M Urea. |
Purity or Quality Grade | Treponema Pallidum protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant other T. pallidum p17 (Partial) Protein >
Spot an opportunity for improvement?Share a Content Correction