missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant P. aeruginosa Lysyl endopeptidase Protein Product Code.: 16062901

enQuireBio™ Recombinant P. aeruginosa Lysyl endopeptidase Protein

Product Code. 16062901
200 μg, 200µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16062901

Brand: enQuireBio™ QP8146ec200ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) Lysyl endopeptidase, was expressed in the hosts and tags indicated. option, ab

Specifications

Accession Number NP_252864.1
Gene ID (Entrez) 880208
Name P. aeruginosa Lysyl endopeptidase Protein
Quantity 200 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol prpL
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity P. aeruginosa
Species E. coli
Protein Tag His-SUMO
Sequence AGYRDGFGAS GSCEVDAVCA TQSGTRAYDN ATAAVAKMVF TSSADGGSYI CTGTLLNNGN SPKRQLFWSA AHCIEDQATA ATLQTIWFYN TTQCYGDAST INQSVTVLTG GANILHRDAK RDTLLLELKR TPPAGVFYQG WSATPIANGS LGHDIHHPRG DAKKYSQGNV SAVGVTYDGH TALTRVDWPS AVVEGGSSGS GLLTVAGDGS YQLRGGLYGG PSYCGAPTSQ RNDYFSDFSG VYSQISRYFA P
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant P. aeruginosa Lysyl endopeptidase Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.