missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. gingivalis Lys-gingipain Protein
A DNA sequence encoding the Porphyromonas gingivalis (strain ATCC 33277 / DSM 20709 / JCM 12257) Lys-gingipain, was expressed in the hosts and tags indicated.
3070.00 SEK - 38570.00 SEK
Specifications
Accession Number | WP_012458488.1 |
---|---|
Gene ID (Entrez) | 29256891 |
Name | P. gingivalis Lys-gingipain Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16068860
|
enQuireBio™
QP7438-EC-100UG |
100 μg |
11125.00 SEK
100µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088860
|
enQuireBio™
QP7438-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078860
|
enQuireBio™
QP7438-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098860
|
enQuireBio™
QP7438-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008870
|
enQuireBio™
QP7438-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018870
|
enQuireBio™
QP7438-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_012458488.1 | |
P. gingivalis Lys-gingipain Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
P. gingivalis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
29256891 | |
Research Use Only | |
kgp | |
Recombinant Protein | |
E. coli | |
DVYTDHGDLY NTPVRMLVVA GAKFKEALKP WLTWKAQKGF YLDVHYTDEA EVGTTNASIK AFIHKKYNDG LAASAAPVFL ALVGDTDVIS GEKGKKTKKV TDLYYSAVDG DYFPEMYTFR MSASSPEELT NIIDKVLMYE KATMPDKSYL EKALLIAGAD SYWNPKIGQQ TIKYAVQYYY NQDHGYTDVY SYPKAPYTGC YSHLNTGVGF ANYTAHGSET SWADPSLTAT QVKALTNKDK YFLAIGNCCV TAQFDYPQPC FGEVMTRVKE KGAYAYIGSS PNSYWGEDYY WSVGANAVFG VQPTFEGTSM GSYDATFLED SYNTVNSIMW AGNLAATHAG NIGNITHIGA HYYWEAYHVL GDGSVM | |
Greater than 82.5% as determined by SDS-PAGE. |