missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit IL17 / IL17A Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) IL17 / IL17A, was expressed in the hosts and tags indicated.
3490.00 SEK - 32610.00 SEK
Specifications
Accession Number | XP_002714544.1 |
---|---|
Gene ID (Entrez) | 100339322 |
Name | IL17 / IL17A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16081610
|
enQuireBio™
QP6212-ye-10ug |
10 ÎĽg | N/A | ||||||
16021620
|
enQuireBio™
QP6212-ye-50ug |
50 ÎĽg | N/A | ||||||
16071610
|
enQuireBio™
QP6212-ye-100ug |
100 ÎĽg |
12935.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16001620
|
enQuireBio™
QP6212-ye-200ug |
200 ÎĽg | N/A | ||||||
16011620
|
enQuireBio™
QP6212-ye-500ug |
500 ÎĽg |
31245.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16091610
|
enQuireBio™
QP6212-ye-1mg |
1 mg | N/A | ||||||
Specifications
XP_002714544.1 | |
IL17 / IL17A Protein | |
Not determined. | |
Full Length | |
Rabbit | |
His | |
Tris-based buffer, 50% glycerol |
100339322 | |
Research Use Only | |
IL17A | |
Recombinant Protein | |
Yeast | |
VKNGIAMPRN PGCPNAEDKN FPQNVKVSLN ILNKSVNSRR PSDYYNRSTS PWTLHRNEDR ERYPSVIWEA KCRHLGCVNA EGNEDHHMNS VPIQQEILVL RRESQHCPHS FRLEKMLVAV GCTCVTPIIH HMA | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Rabbit IL17 / IL17A Protein