missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Anionic trypsin-1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Anionic trypsin-1, was expressed in the hosts and tags indicated.
2430.00 SEK - 26260.00 SEK
Specifications
Accession Number | NP_036767.1 |
---|---|
Gene ID (Entrez) | 24691 |
Name | Anionic trypsin-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16098141
|
enQuireBio™
QP9349-ye-10ug |
10 ÎĽg | N/A | ||||||
16038151
|
enQuireBio™
QP9349-ye-50ug |
50 ÎĽg | N/A | ||||||
16088141
|
enQuireBio™
QP9349-ye-100ug |
100 ÎĽg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16018151
|
enQuireBio™
QP9349-ye-200ug |
200 ÎĽg | N/A | ||||||
16028151
|
enQuireBio™
QP9349-ye-500ug |
500 ÎĽg |
25885.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16008151
|
enQuireBio™
QP9349-ye-1mg |
1 mg | N/A | ||||||
Specifications
NP_036767.1 | |
Anionic trypsin-1 Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
24691 | |
Research Use Only | |
Prss1 | |
Recombinant Protein | |
Yeast | |
IVGGYTCPEH SVPYQVSLNS GYHFCGGSLI NDQWVVSAAH CYKSRIQVRL GEHNINVLEG DEQFINAAKI IKHPNYSSWT LNNDIMLIKL SSPVKLNARV APVALPSACA PAGTQCLISG WGNTLSNGVN NPDLLQCVDA PVLSQADCEA AYPGEITSSM ICVGFLEGGK DSCQGDSGGP VVCNGQLQGI VSWGYGCALP | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Rat Anionic trypsin-1 Protein