missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat MAPT / Tau Protein
A DNA sequence encoding the Rattus norvegicus (Rat) MAPT / Tau, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6345-ec-50ug
This item is not returnable.
View return policy
Specifications
MAPT / Tau Protein | |
Research Use Only | |
Mapt | |
Recombinant Protein | |
E. coli | |
AEPRQEFDTM EDQAGDYTML QDQEGDMDHG LKESPPQPPA DDGSEEPGSE TSDAKSTPTA EDVTAPLVEE RAPDKQATAQ SHTEIPEGTT AEEAGIGDTP NMEDQAAGHV TQEPQKVEIF SQSLLVEPGR REGQAPDSGI SDWTHQQVPS MSGAPLPPQG LREATHQPLG TRPEDVERSH PASELLWQES PQKEAWGKDR LGSEEEVDED ITMDESSQES PPSQASLAPG TATPQARSVS ASGVSGETTS IPGFPAEGSI PLPADFFSKV SAETQASPPE GPGTGPSEEG HEAAPEFTFH VEIKASAPKE QDLEGATVVG APAEEQKARG PSVGKGTKEA SLLEPTDKQP AAGLPGRPVS RVPQLKARVA GVSKDRTGND EKKAKTSTPS CAKTPSNRPC LSPTRPTPGS SDPLIKPSSP AVCPEPATSP KYVSSVTPRN GSPGTKQMKL KGADGKTGAK IATPRGAATP GQKGTSNATR IPAKTTPSPK TPPGSGEPPK SGERSGYSSP GSPGTPGSRS RTPSLPTPPT REPKKVAVVR TPPKSPSASK SRLQTAPVPM PDLKNVRSKI GSTENLKHQP GGGKVQIINK KLDLSNVQSK CGSKDNIKHV PGGGSVHIVY KPVDLSKVTS KCGSLGNIHH KPGGGQVEVK SEKLDFKDRV QSKIGSLDNI THVPGGGNKK IETHKLTFRE NAKAKTDHGA EIVYKSPVVS GDTSPRHLSN VSSTGSIDMV DSPQLATLAD EVSASLAKQG L | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Rat MAPT / Tau Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction