missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PTMA Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PTMA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9354-ye-10ug
This item is not returnable.
View return policy
Specifications
NP_068508.1 | |
PTMA Protein | |
Research Use Only | |
Ptma | |
Recombinant Protein | |
Yeast | |
SDAAVDTSSE ITTKDLKEKK EVVEEAENGR DAPANGNAQN EENGEQEADN EVDEEEEEGG EEEEEEEEGD GEEEDGDEDE EAEAPTGKRV AEDDEDDDVE TKKQKKTDED D | |
Greater than 90% as determined by SDS-PAGE. |
100359583 | |
10 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Rat PTMA Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction